Learn More
Isoamyl acetate, 99+%, pure, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 123-92-2 |
---|---|
Molecular Formula | C7H14O2 |
Molecular Weight (g/mol) | 130.19 |
MDL Number | MFCD00008946 |
InChI Key | MLFHJEHSLIIPHL-UHFFFAOYSA-N |
Synonym | isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate |
PubChem CID | 31276 |
ChEBI | CHEBI:31725 |
IUPAC Name | 3-methylbutyl acetate |
SMILES | CC(C)CCOC(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10103250
|
Thermo Scientific Acros
150662500 |
250 mL | Glass bottle |
N/A
|
N/A | ||||
10655582
|
Thermo Scientific Acros
150660010 |
1 L | Glass bottle |
N/A
|
N/A | ||||
10174473
|
Thermo Scientific
150660025 |
2.5 L | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-92-2 | |
130.19 | |
MLFHJEHSLIIPHL-UHFFFAOYSA-N | |
31276 | |
3-methylbutyl acetate |
C7H14O2 | |
MFCD00008946 | |
isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate | |
CHEBI:31725 | |
CC(C)CCOC(C)=O |
Specifications
123-92-2 | |
0.8740g/mL | |
25°C | |
C7H14O2 | |
1.3990 to 1.4010 | |
02, 132 | |
0.874 | |
Solubility in water: 2mg/mL (20°C). Other solubilities: soluble in ethanol,diethyl phthalate,mineral oil,and 1,2-propanediol | |
CC(C)CCOC(C)=O | |
130.19 | |
CHEBI:31725 | |
130.19 | |
Pure |
-78.0°C | |
142.0°C (756.0 mmHg) | |
99% min. (GC) | |
CH3CO2CH2CH2CH(CH3)2 | |
MFCD00008946 | |
15, 5156 | |
isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate | |
MLFHJEHSLIIPHL-UHFFFAOYSA-N | |
3-methylbutyl acetate | |
31276 | |
1.03 Pas (8.97°C) | |
99+% | |
Isoamyl acetate |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-662-3
RUO – Research Use Only