Learn More
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
£24.00 - £50.50
Chemical Identifiers
CAS | 79-31-2 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.11 |
MDL Number | MFCD00002658 |
InChI Key | KQNPFQTWMSNSAP-UHFFFAOYSA-N |
Synonym | isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl |
PubChem CID | 6590 |
ChEBI | CHEBI:16135 |
IUPAC Name | 2-methylpropanoic acid |
SMILES | CC(C)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10173210
|
Thermo Scientific Acros
122520250 |
25 g | Glass bottle |
£24.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10041170
|
Thermo Scientific Acros
122525000 |
500 g | Glass bottle |
£33.40
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10356470
|
Thermo Scientific Acros
122520010 |
1 kg | Glass bottle |
£50.50
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagentChemical Identifiers
79-31-2 | |
88.11 | |
KQNPFQTWMSNSAP-UHFFFAOYSA-N | |
6590 | |
2-methylpropanoic acid |
C4H8O2 | |
MFCD00002658 | |
isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl | |
CHEBI:16135 | |
CC(C)C(O)=O |
Specifications
79-31-2 | |
153.0°C to 154.0°C | |
99+% | |
(CH3)2CHCO2H | |
02, 288 | |
isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl | |
KQNPFQTWMSNSAP-UHFFFAOYSA-N | |
2-methylpropanoic acid | |
6590 | |
88.11 | |
0.9500g/mL | |
Authentic | |
1.3920 to 1.3940 | |
Isobutyric acid, 99+% |
-47.0°C | |
Colorless | |
C4H8O2 | |
MFCD00002658 | |
15, 5200 | |
Solubility in water: 210g/L (20°C). Other solubilities: miscible with alcohol, chloroform and ether | |
CC(C)C(O)=O | |
88.11 | |
CHEBI:16135 | |
≥99% | |
59°C | |
Liquid | |
0.95 |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-195-7
missing translation for 'rtecsNumber' : NQ4375000
missing translation for 'tsca' : TSCA
- Isobutyric acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only