missing translation for 'onlineSavingsMsg'
Learn More

Isobutyronitrile, 99%, Thermo Scientific Chemicals

£27.90 - £60.20

Chemical Identifiers

CAS 78-82-0
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00001873
InChI Key LRDFRRGEGBBSRN-UHFFFAOYSA-N
Synonym isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid
PubChem CID 6559
ChEBI CHEBI:28638
IUPAC Name 2-methylpropanenitrile
SMILES CC(C)C#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11393526
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L14470.AK
250 mL
£27.90
250mL
Please to purchase this item. Need a web account? Register with us today!
11303536
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L14470.AU
1000 mL
£60.20
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

Solubility
Soluble in water 35g/L (20°C).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

78-82-0
69.107
LRDFRRGEGBBSRN-UHFFFAOYSA-N
6559
2-methylpropanenitrile
C4H7N
MFCD00001873
isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid
CHEBI:28638
CC(C)C#N

Specifications

78-82-0
0.77
8°C (46°F)
(CH3)2CHCN
MFCD00001873
1340512
isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid
LRDFRRGEGBBSRN-UHFFFAOYSA-N
2-methylpropanenitrile
6559
69.11
Isobutyronitrile
-72°C
103°C to 104°C
C4H7N
1.374
UN2284
14,5156
Soluble in water 35g/L (20°C).
CC(C)C#N
69.107
CHEBI:28638
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H311-H330
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P309+P311-P310-P312-P330-P363-P370+P378q-P501c

H225-H300+H330-H311-H371

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYRONITRILE

missing translation for 'einecsNumber' : 201-147-5

missing translation for 'rtecsNumber' : TZ4900000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Isobutyronitrile

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Isobutyronitrile

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only