missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Isolator Wrap Sterile Settle Tryptic Soy Agar Plates
Thermo Scientific™ Remel Sterile Settling Plate Tryptic Soy Agar is used for the cultivation of bacteria, yeast and fungi.
Brand: Thermo Scientific™ R111816
79.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- VHP resistant packaged Settling plates (single bag and double bag) are manufactured in a clean room environment. Irradiated with test indicators. This provides the highest level of sterility assurance with no compromise of growth characteristics.
- Each lot is tested to ensure consistent, reproducible results.
- Storage Condition: 2 to 8°C.

Specifications
2°C to 8°C, Refrigerated | |
Isolator Settling Plate | |
10/Pk. |
Tryptic Soy Agar | |
RABS, Isolators and Cleanrooms | |
85 mm |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
242712 | Certificate of Analysis | 27/03/2025 | R111816 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Isolator Wrap Sterile Settle Tryptic Soy Agar Plates > 85 mm; Isolator Sterile Settle Tryptic Soy Agar Plates; 10/Pk.
Spot an opportunity for improvement?Share a Content Correction