missing translation for 'onlineSavingsMsg'
Learn More

Merck Isopore™ Membrane Filters, 0.1 μm

Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.

Brand:  Merck VCTP02500

Code : 71

Additional Details : Weight : 1.81440kg

 View more versions of this product

Product Code. 10695761

  • £321.00 / Pack of 100
Estimated Shipment: 20-08-2024
to see stock.
Add to basket

Description

Description

Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.

The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.

Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety

Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays

Specifications

Specifications

Not Autoclavable
White
Polycarbonate
Non-sterile
Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence.
Hydrophilic
100/Pk.
Hydrophilic
25 mm
101 psi
Screen Filter
0.1 μm
Avg. 2.5 mL/min x cm2 (typical results (at 10 psi);0.13 mL/min x cm2/psi)
1.5 L/min/cm2
100Pack
Filter Membranes
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers