Learn More
Merck Isopore™ Membrane Filters, 0.1 μm
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
£321.00 - £890.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | White |
Filter Type | Screen Filter |
Pore Size | 0.1 μm |
Sterility | Non-sterile |
Product Code | Brand | Diameter (Metric) | Bubble Point | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Bubble Point | Price | Quantity & Availability | ||||
10106380
|
Merck
VCTP01300 |
13 mm | 101 psi |
£322.00
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||||
10695761
|
Merck
VCTP02500 |
25 mm | 101 psi |
£321.00
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||||
10656151
|
Merck
VCTP04700 |
47 mm | 101 psi |
£490.20
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||||
10409532
|
Merck
VCTP14250 |
142 mm | 7.0 bar, air with water |
£890.00
Pack of 50 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||||
Description
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
Not Autoclavable | |
Screen Filter | |
Non-sterile | |
Hydrophilic |
White | |
0.1 μm | |
Filter Membranes |