missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, Certified AR for Analysis, Fisher Chemical™

C3H8O, CAS Number-67-63-0, 2-hydroxypropane, isopropanol, hartosol, 2-propanol, dimethylcarbinol, avantine, petrohol, sec-propyl alcohol, isopropyl alcohol, alkolave, 2.5L, 81 deg.C, CHEBI:17824, Colorless, 60.1g/mol, KFZMGEQAYNKOFK-UHFFFAOYSA-N, propan-2-ol, -89.5 deg.C, 11674, 60.096

£66.60 - £839.00

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number 11674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 9
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11453733
View Documents
Fisher Chemical
P/7500/PB08
500 mL Plastic-coated Glass Bottle
£77.95
500mL
Please to purchase this item. Need a web account? Register with us today!
10366430
View accessory products View Documents
Fisher Chemical
P/7500/15
1 L Amber Glass Bottle
£66.60
1 L
Please to purchase this item. Need a web account? Register with us today!
10315720
View accessory products View Documents Promotion Details
Fisher Chemical
P/7500/17
2.5 L Amber Glass Bottle
£145.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10628143
View Documents Promotion Details
Fisher Chemical
P/7500/PC17
2.5 L HDPE Bottle
£158.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10497070
View Documents
Fisher Chemical
P/7500/PB17
2.5 L Plastic-coated Glass Bottle
£161.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10080320
View Documents Promotion Details
Fisher Chemical
P/7500/21
5 L HDPE Bottle
£245.00
5 L
Please to purchase this item. Need a web account? Register with us today!
10522992
View Documents
Fisher Chemical
P/7500/24
10 L Plastic Container
£492.00
10 L
Please to purchase this item. Need a web account? Register with us today!
10090320
View Documents
Fisher Chemical
P/7500/25
25 L Metal Can
£717.00
25 L
Please to purchase this item. Need a web account? Register with us today!
12656757
View Documents
Fisher Chemical
P/7500/PC25
25 L Plastic Container
£839.00
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C3H8O, CAS Number-67-63-0, 2-hydroxypropane, isopropanol, hartosol, 2-propanol, dimethylcarbinol, avantine, petrohol, sec-propyl alcohol, isopropyl alcohol, alkolave, 2.5L, 81 deg.C, CHEBI:17824, Colorless, 60.1g/mol, KFZMGEQAYNKOFK-UHFFFAOYSA-N, propan-2-ol, -89.5 deg.C, 11674, 60.096

Specifications

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
11674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O

Specifications

7
67-63-0
81°C
C3H8O
43mmHg at 20°C
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
Isopropanol
-89.5°C
Colorless
Liquid
11674
2.27 mPaS at 20°C
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Propan-2-ol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P240-Ground/bond container and receiving equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.