Learn More
Isopropenyl acetate, 99%, Thermo Scientific Chemicals
£24.20 - £69.80
Chemical Identifiers
CAS | 108-22-5 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00008709 |
InChI Key | HETCEOQFVDFGSY-UHFFFAOYSA-N |
Synonym | isopropenyl acetate, 2-acetoxypropene, 1-methylvinyl acetate, 2-acetoxypropylene, methylvinyl acetate, 1-propen-2-ol, acetate, propen-2-yl acetate, acetic acid isopropenyl ester, 1-acetoxy-1-methylethylene, 1-propen-2-yl acetate |
PubChem CID | 7916 |
IUPAC Name | prop-1-en-2-yl acetate |
SMILES | CC(=C)OC(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10357900
|
Thermo Scientific Acros
152502500 |
250 g | Glass bottle |
£24.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10337950
|
Thermo Scientific Acros
152500010 |
1 kg | Glass bottle |
£69.80
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-22-5 | |
100.12 | |
HETCEOQFVDFGSY-UHFFFAOYSA-N | |
7916 | |
CC(=C)OC(C)=O |
C5H8O2 | |
MFCD00008709 | |
isopropenyl acetate, 2-acetoxypropene, 1-methylvinyl acetate, 2-acetoxypropylene, methylvinyl acetate, 1-propen-2-ol, acetate, propen-2-yl acetate, acetic acid isopropenyl ester, 1-acetoxy-1-methylethylene, 1-propen-2-yl acetate | |
prop-1-en-2-yl acetate |
Specifications
108-22-5 | |
Colorless to Yellow | |
94°C to 97°C | |
Authentic | |
C5H8O2 | |
1.4000 to 1.402 | |
02, II, 278 | |
15, 5249 | |
isopropenyl acetate, 2-acetoxypropene, 1-methylvinyl acetate, 2-acetoxypropylene, methylvinyl acetate, 1-propen-2-ol, acetate, propen-2-yl acetate, acetic acid isopropenyl ester, 1-acetoxy-1-methylethylene, 1-propen-2-yl acetate | |
HETCEOQFVDFGSY-UHFFFAOYSA-N | |
prop-1-en-2-yl acetate | |
7916 | |
99% | |
Isopropenyl acetate, 99% |
-93°C | |
0.9200g/mL | |
10°C | |
98.5% min. (GC) | |
CH3CO2C(CH3)=CH2 | |
MFCD00008709 | |
01,524; 04,273; 06,308 | |
0.92 | |
Solubility in water: 34g/L in water (20°C). Other solubilities: miscible with nearly all organic solvents | |
CC(=C)OC(C)=O | |
100.12 | |
100.12 | |
Liquid |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-562-7
missing translation for 'rtecsNumber' : UD4200000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.