missing translation for 'onlineSavingsMsg'
Learn More
Isopropylmagnesium chloride, 2.0M solution in THF, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10726243

Isopropylmagnesium chloride, 2.0M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10726243
800 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10726243

Brand: Thermo Scientific Acros 212858000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-55-9, 109-99-9
Molecular Formula C3H7ClMg
Molecular Weight (g/mol) 102.84
MDL Number MFCD00000466
InChI Key GPIBKUJXKCEZOH-UHFFFAOYSA-M
Synonym isopropylmagnesium chloride, magnesium, chloro 1-methylethyl, chloro 1-methylethyl magnesium, i-prmgcl, isopropylmagnesiumchloride, chloro isopropyl magnesium, isopropyl magnesium chloride, isopropylmagnesium chloride solution, iprmgcl, ipr-mgclShow MoreShow Less
PubChem CID 101955
SMILES CC(C)[Mg]Cl

Specifications

Color Brown to Brown-Yellow
Boiling Point 62.4°C (899.8hPa)
Melting Point 5.0°C
Linear Formula (CH3)2CHMgCl
Concentration 1.80 to 2.20M
Density 0.9700g/mL
Solubility Information Solubility in water: vigorous reaction.
IUPAC Name chloro(propan-2-yl)magnesium
Formula Weight 102.85
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.97
Quantity 800 mL
Flash Point −27°C
Physical Form Solution
Chemical Name or Material Isopropylmagnesium chloride, 2.0M Solution in THF
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0454965 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number212858000,212851000
Lot NumberA0454966 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number212858000,212851000
Lot NumberA0454964 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number212858000,212851000
Lot NumberA0450486 Certificate TypeCertificate of Analysis Date13/09/2023 Catalog Number212858000,212851000
Lot NumberA0454966 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number212858000,212851000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isopropylmagnesium chloride, 2.0M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    30

    Hours

    5

    Minutes

    45

    Seconds

    41

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isopropylmagnesium chloride, 2.0M solution in THF, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.