missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 6200 H11
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Brand: Fisherbrand™ 211312100
1336.79 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
Standard high temperature cutout (HTC) circuitry is user-selectable to ensure application temperatures do not exceed set point parameters- High temperature and low level cut-outs
- Compact footprint maximizes bench space
- Includes external circulation plumbing with 8-mm and 12-mm fittings and supply and return clamps
- Work area cover included on all heated baths
- High quality stainless steel baths
- All bath drains are located on the front of the unit with an integrated valve
Specifications
5.1 to 6.5 L | |
10°C to 200°C | |
6200 H7 | |
10. kg | |
230 V |
Force and Suction | |
2000 w | |
36.32 x 21.59 x 414 cm | |
230 V 50 Hz | |
50 Hz |