Learn More
Eppendorf™ IsoTherm™ System Starter Set
Holds up to 24 micro tubes. The IsoTherm-System is the ideal system for safely cooling your samples in 0.5mL or 1.5 mL/2.0 mL tube formats at -21°C or 0°C throughout your entire lab workflow.
£158.55 - £167.67
Specifications
Type IsoTherm-System Starter Set
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10114721
View Documents
Eppendorf™
3880000011
0.5mL tubes N/A N/A N/A
10160231
View Documents Promotion Details
Eppendorf™
3880001018
1,5/2,0mL tubes
£167.67
Each
In Stock
to see stock.
Add to basket
Description
Description
  • IsoPack™ (cold pack) white for 0°C and blue for -21°C, cooled to refrigeration temperature in freezer compartment
  • Isothermal times of >6hr (0°C, white) and >3hr (-21°C, blue)
  • No more melting ice baths (danger of contamination)
  • All system components can be used separately
  • Racks are stackable, autoclavable, markable, floatable and can be centrifuged in a microtitre-plate rotor
  • Specializes in cooling samples effectively and constantly at -21°C or 0°C over many hours.
Specifications
Specifications
IsoTherm-System Starter Set
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Eppendorf™ IsoTherm™ System Starter Set