Learn More
CellSensor™ ISRE-bla HEK293T Cell Line
CellSensor™ ISRE-bla HEK293T Cell Line is responsive to interferon alpha and can be used to probe the JAK/STAT signaling pathway.
Brand:  CellSensor™ K1655
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 10563084
  • £7620.00 / Each
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
Description
Description
  • Interferon cell signaling is a critical pathway involved in viral defense and autoimmune disease.
  • The CellSensor™ ISRE-bla HEK 293T cell line contains the β-lactamase reporter gene under the control of the interferon stimulated response element (ISRE).
  • This cell line was engineered by transduction of the ISRE-bla construct into HEK 293T cells by lentivirus.
  • Flow cytometry was then used to isolate a pool of clones responsive to interferon alpha a stimulation.
  • A dose response curve for this assay against interferon alpha is available in the validation packet.
Specifications
Specifications
Store in liquid nitrogen immediately upon receipt, or thaw for immediate use
Cytokines, Signaling Pathway
1 Vial
Dividing Cells