Learn More
Tocris Bioscience™ J 147
Neuroprotective and neurotrophic compound; reduces Aβ40 and Aβ42 levels
Brand:  Tocris Bioscience™ 5081/10
 View more versions of this product
Product Code. 18793429
  • £179.00 / 10mg
Explore more special offers
Description
Description
J 147 is a potent neuroprotective and neurotrophic compound. Protects against neurotoxicity in cortical neurons in vitro (EC50 = 25 - 200 nM). Reduces soluble Aβ40 and Aβ42 levels and increases BDNF levels in the hippocampus in vivo. Enhances memory in wild type rats. Prevents memory deficits and rescues cognitive deficits in young and aged transgenic Alzheimer's disease mice, respectively. Orally active.
Specifications
Specifications
>99%
J 147
J 147
10 mg
350.33