missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ J 147

Neuroprotective and neurotrophic compound; reduces Aβ40 and Aβ42 levels

£179.00 - £658.00

Specifications

Purity >99%
Product Type J 147
Molecular Weight (g/mol) 350.33
Inhibitors J 147
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18793429
View Documents
Tocris Bioscience™
5081/10
10 mg
£179.00
10mg
Please to purchase this item. Need a web account? Register with us today!
18769716
View Documents
Tocris Bioscience™
5081/50
50 mg
£658.00
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent neuroprotective and neurotrophic compound. Protects against neurotoxicity in cortical neurons in vitro (EC50 = 25 - 200 nM). Reduces soluble Aβ40 and Aβ42 levels and increases BDNF levels in the hippocampus in vivo. Enhances memory in wild type rats. Prevents memory deficits and rescues cognitive deficits in young and aged transgenic Alzheimer's disease mice, respectively. Orally active.
Specifications

Specifications

>99%
J 147
350.33
J 147
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.