Learn More
GPE Scientific™ J Young NMR Valve with Tube Attached
GPE Scientific™ J Young NMR Valve with Tube Attached allows you to handle your NMR sample without flame sealing your tubes.
Brand:  GPE Scientific™ S-5-500-JY-7-YELLOW
 View more versions of this product
Product Code. 15874874
  • £160.00 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Handle your NMR sample without flame sealing your tubes.
  • Teflon covered O-ring eliminates material incompatibilities.
  • Completely greaseless Teflon assembly means the tube is easy to use and to disassemble for cleaning.
  • Includes female joint for quick attachment to your vacuum rack.
  • Yellow piston
  • Negative pressure/high vacuum.
  • Leak tested to 10-9mbar liters/sec.
  • Internal positive pressure 5bar, 72psi
Specifications
Specifications
500 MHz
5mm NMR Valve
7 in.