Learn More
GPE Scientific™ J Young O-Rings for Spherical Joints
GPE Scientific™ J Young O-Rings for Spherical Joints facilitate a secure joint seal for spherical joints and are available as Viton or PTFE-coated Viton.
Brand: GPE Scientific™ O51/P
Additional Details : Weight : 0.00100kg
Description
- Suitable for use with spherical joints
- Available in Viton or PTFE-coated Viton
- Available in different joint sizes
Specifications
O-ring for Spherical Joint | |
PTFE-Covered Viton | |
51/35 size spherical joints |