Learn More
GPE Scientific™ J Young O-Rings for Spherical Joints
GPE Scientific™ J Young O-Rings for Spherical Joints facilitate a secure joint seal for spherical joints and are available as Viton or PTFE-coated Viton.
£1.00 - £118.00
Product Code | Brand | Material | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Size | Price | Quantity & Availability | ||||
15824814
|
GPE Scientific™
O12/P |
PTFE-Covered Viton | 12/2 and 12/5 size spherical joints |
£31.35
Each |
Estimated Shipment: 19-09-2024
Log in to see stock. |
||||
15834814
|
GPE Scientific™
O12/V |
Viton | 12/2 and 12/5 size spherical joints |
£1.00
Each |
Estimated Shipment: 19-09-2024
Log in to see stock. |
||||
15804824
|
GPE Scientific™
O40/P |
PTFE-Covered Viton | 40/25 size spherical joints |
£101.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||||
15854814
|
GPE Scientific™
O18/V |
Viton | 18/7 and 18/9 size spherical joints |
£1.00
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
15864814
|
GPE Scientific™
O28/P |
PTFE-Covered Viton | 28/12 and 28/15 size spherical joints |
£71.25
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||||
15874814
|
GPE Scientific™
O28/V |
Viton | 28/12 and 28/15 size spherical joints |
£1.20
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
15884814
|
GPE Scientific™
O35/P |
PTFE-Covered Viton | 35/20 size spherical joints |
£105.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||||
15894814
|
GPE Scientific™
O35/V |
Viton | 35/20 size spherical joints |
£1.40
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
15844814
|
GPE Scientific™
O18/P |
PTFE-Covered Viton | 18/7 and 18/9 size spherical joints |
£47.50
Each |
Estimated Shipment: 19-09-2024
Log in to see stock. |
||||
15814824
|
GPE Scientific™
O40/V |
Viton | 40/25 size spherical joints |
£1.50
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
15824824
|
GPE Scientific™
O51/P |
PTFE-Covered Viton | 51/35 size spherical joints |
£118.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||||
15834824
|
GPE Scientific™
O51/V |
Viton | 51/35 size spherical joints |
£2.05
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
Description
- Suitable for use with spherical joints
- Available in Viton or PTFE-coated Viton
- Available in different joint sizes
Specifications
O-ring for Spherical Joint |