Learn More
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Fine Thread Tap and Socket
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Fine Thread Tap and Socket is specially designed for use with Schlenk lines and vacuum manifolds.
Brand: GPE Scientific™ JRF/100
This item is not returnable.
View return policy
Description
- Solidly constructed for use with vacuum manifolds
- Designed for use with Schlenk lines
- Includes fine thread tap and socket
Specifications
100 mL | |
Fine thread tap and socket | |
Round |
Schlenk lines and vacuum manifolds | |
Size: 24 |