Learn More
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Fine Thread Tap and Socket
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Fine Thread Tap and Socket is specially designed for use with Schlenk lines and vacuum manifolds.
Brand:  GPE Scientific™ JRF/500
 View more versions of this product
Product Code. 15884804
  • £147.00 / Each
Estimated Shipment: 17-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Solidly constructed for use with vacuum manifolds
  • Designed for use with Schlenk lines
  • Includes fine thread tap and socket
Specifications
Specifications
500 mL
Fine thread tap and socket
Round
Schlenk lines and vacuum manifolds
Size: 24