missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Janus Green B, pure, high purity biological stain
£60.40 - £202.00
Chemical Identifiers
CAS | 2869-83-2 |
---|---|
Molecular Formula | C30H31ClN6 |
Molecular Weight (g/mol) | 511.07 |
MDL Number | MFCD00011758 |
InChI Key | XXACTDWGHQXLGW-UHFFFAOYSA-M |
Synonym | C.I. 11050 |
PubChem CID | 76123 |
ChEBI | CHEBI:21184 |
IUPAC Name | 8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride |
SMILES | [Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10163730
|
Thermo Scientific Chemicals
191680050 |
5 g | Glass bottle |
£60.40
5g |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
10340644
|
Thermo Scientific Chemicals
191680250 |
25 g | Glass bottle |
£202.00
25g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2869-83-2 | |
511.07 | |
XXACTDWGHQXLGW-UHFFFAOYSA-M | |
76123 | |
8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride |
C30H31ClN6 | |
MFCD00011758 | |
C.I. 11050 | |
CHEBI:21184 | |
[Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C |
Specifications
2869-83-2 | |
C30H31ClN6 | |
15, 5303 | |
Solubility in water: soluble. | |
Authentic | |
8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride | |
76123 | |
511.07 | |
Black or Brown/Green | |
Janus Green B, Pure |
50% min. | |
MFCD00011758 | |
C.I. 11050 | |
XXACTDWGHQXLGW-UHFFFAOYSA-M | |
[Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C | |
511.07 | |
CHEBI:21184 | |
Pure | |
Powder |
RUO – Research Use Only