missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Jenway™ 100 Disposable Cuvettes

Brand:  Cole-Parmer™ 83070-32

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11420559

  • £102.61 / Pack of 100
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Cuvette
2.4 to 4.5 mL
100/Pk.
Spectral Measurement of Liquids
UV and visible wavelengths
Plastic
10 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers