missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jenway™ Conductivity Standards
Determines a sample's electrical conductivity with Bibby Scientific™ Jenway™ Conductivity Standards provide stable and reliable calibrations.
£68.30 - £78.50
Product Code | Brand | Quantity | Conductivity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Conductivity | Price | Quantity & Availability | ||||
11400509
|
Jenway
16156-80 |
500 mL | 84 μS |
£78.50
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11490499
|
Jenway
90238-48 |
500 mL | 12.88 mS |
£68.30
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For use with Jenway™ Conductivity and TDS meters.
- Choose 12.88m/S or 84μ/S standards depending on your specific needs
- Available in quantities of 500mL
- For use with Jenway™ Conductivity and TDS meters.
- Choose 12.88m/S or 84μ/S standards depending on your specific needs
- Available in quantities of 500mL
Chemical Identifiers
500 mL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title