Learn More
Cole-Parmer™ Jenway™ Epoxy Bodied Conductivity Cells with DIN Connector
Brand:  Cole-Parmer™ 99959-66
229.02 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 13127624
  • £335.00 / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.1
Conductivity Cell
Epoxy
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Jenway™ Epoxy Bodied Conductivity Cells with DIN Connector > Cell Constant: 0.1