Learn More
Jenway™ pH Electrodes
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
Brand: Jenway™ 90269-17
This item is not returnable.
View return policy
Description
- Quickly reach equilibrium
- Significantly reduced response times
- Far greater reproducibility
- Significantly reduced long-term drift
Specifications
BNC | |
Refillable Spear Type Combination with Bulb End | |
measurements in small volumes and soil and slurry samples | |
Glass | |
0°C to 80°C |
6 mm | |
6 dia. x 40 mm L | |
40 mm | |
0 to 14 |