Learn More
Jenway™ pH Electrodes
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
Brand:  Jenway™ 99960-41
 View more versions of this product
Product Code. 13107184
  • £164.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift
Specifications
Specifications
BNC
Life Science Refillable Combination with Bulb End
measurement of samples in test tubes and Eppendorf vessels
Glass
0°C to 80°C
6 mm
6 dia. x 150 mm L
150 mm
0 to 14