missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JTV 519 fumarate
Ryanodine receptor (RyR) inhibitor
Brand: Tocris Bioscience™ 4564/10
This item is not returnable.
View return policy
Description
JTV 519 fumarate is a ryanodine receptor (RyR) inhibitor; stabilizes RyR2 in a closed conformation. Exhibits antiarrhythmic and cardioprotective properties in vivo.
Specifications
98% | |
JTV 519 fumarate | |
M.W. (Observed): 540.67 | |
10 mg |
Store at +4°C | |
K 201 | |
JTV 519 fumarate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ JTV 519 fumarate > 10mg
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction