missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JTV 519 fumarate
Ryanodine receptor (RyR) inhibitor
£130.00 - £491.00
Specifications
Purity | 98% |
---|---|
Content And Storage | Store at +4°C |
Description | JTV 519 fumarate |
Gene Alias | K 201 |
Molecular Weight (g/mol) | M.W. (Observed): 540.67 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18758808
|
Tocris Bioscience™
4564/10 |
10 mg |
£130.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18785668
|
Tocris Bioscience™
4564/50 |
50 mg |
£491.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ryanodine receptor (RyR) inhibitor; stabilizes RyR2 in a closed conformation. Exhibits antiarrhythmic and cardioprotective properties in vivo.Specifications
98% | |
JTV 519 fumarate | |
M.W. (Observed): 540.67 |
Store at +4°C | |
K 201 | |
JTV 519 fumarate |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ JTV 519 fumarate