missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Jumbosep™ Centrifugal Device Membrane Inserts
Jumbosep™ devices can accelerate sample processing, maximize sample recovery, add versatility, and prevent solution bypass.
Brand: Cytiva (formerly Pall Lab) OD010C65
Additional Details : Weight : 0.19500kg
Description
- Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes
- Achieve 50X concentration and > 90% recovery in just minutes
- Low protein-binding Omega™ membrane and polysulfone housing minimize losses due to non-specific binding
- Versatile Omega membrane is available in a variety of MWCOs
- Color-coded and laser etched for easy identification
- Built-in dead stop prevents spinning to dryness
- Unique sealing mechanism prevents retentate leakage and filtrate contamination
- Economical - Sample reservoir and filtrate receiver can be sanitized or autoclaved, and reused
Specifications
Blue | |
PES | |
Non-sterile | |
20 to 60 mL (Sample) |
EFA: 15.2 cm2 | |
10 kDa | |
12/Pk. |