missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Jumbosep™ Centrifugal Device Membrane Inserts
Jumbosep™ devices can accelerate sample processing, maximize sample recovery, add versatility, and prevent solution bypass.
£218.00 - £222.00
Specifications
Description | EFA: 15.2 cm2 |
---|---|
Material (Membrane) | PES |
Sterility | Non-sterile |
Quantity | 12/Pk. |
Volume (Metric) | 20 to 60 mL (Sample) |
Product Code | Brand | MWCO | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Color | Price | Quantity & Availability | ||||
17164781
|
Cytiva (formerly Pall Lab)
OD010C65 |
10 kDa | Blue |
£218.00
Pack of 12 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17104791
|
Cytiva (formerly Pall Lab)
OD030C65 |
30 kDa | Red |
£218.00
Pack of 12 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17144791
|
Cytiva (formerly Pall Lab)
OD100C65 |
100 kDa | Clear |
£218.00
Pack of 12 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
17124781
|
Cytiva (formerly Pall Lab)
OD003C65 |
3 kDa | Gray |
£218.00
Pack of 12 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
17184791
|
Cytiva (formerly Pall Lab)
OD300C65 |
300 kDa | Orange |
£222.00
Pack of 12 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
Description
- Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes
- Achieve 50X concentration and > 90% recovery in just minutes
- Low protein-binding Omega™ membrane and polysulfone housing minimize losses due to non-specific binding
- Versatile Omega membrane is available in a variety of MWCOs
- Color-coded and laser etched for easy identification
- Built-in dead stop prevents spinning to dryness
- Unique sealing mechanism prevents retentate leakage and filtrate contamination
- Economical - Sample reservoir and filtrate receiver can be sanitized or autoclaved, and reused
Specifications
EFA: 15.2 cm2 | |
Non-sterile | |
20 to 60 mL (Sample) |
PES | |
12/Pk. |