missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Jumbosep™ Centrifugal Device Starter Kits
Jumbosep™ Centrifugal Device Starter Kit replaces dialysis, chemical precipitation, and lyophilization.
Brand: Cytiva (formerly Pall Lab) FD010K65
Informations supplémentaires : Weight : 0.72200kg
Description
- Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes
- Provides high recoveries, typically > 90%
- For research use only
Spécification
Blue | |
PES | |
4/Pk. | |
20 to 60 mL (Sample) |
EFA: 15.2 cm2 | |
Non-sterile | |
10 kDa Starter Kit |