missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Jumbosep™ Centrifugal Device Starter Kits
Jumbosep™ Centrifugal Device Starter Kit replaces dialysis, chemical precipitation, and lyophilization.
£174.00 - £204.00
Specifications
Description | EFA: 15.2 cm2 |
---|---|
Sterility | Non-sterile |
Quantity | 4/Pk. |
Volume (Metric) | 20 to 60 mL (Sample) |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
17124721
|
Cytiva (formerly Pall Lab)
FD000K65 |
Generic Starter Kit |
£174.00
Pack of 4 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
17144721
|
Cytiva (formerly Pall Lab)
FD003K65 |
3 kDa Starter Kit |
£204.00
Pack of 4 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
17164721
|
Cytiva (formerly Pall Lab)
FD010K65 |
10 kDa Starter Kit |
£204.00
Pack of 4 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
17174721
|
Cytiva (formerly Pall Lab)
FD030K65 |
30 kDa Starter Kit |
£204.00
Pack of 4 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
17184721
|
Cytiva (formerly Pall Lab)
FD100K65 |
100 kDa Starter Kit |
£201.00
Pack of 4 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
17194721
|
Cytiva (formerly Pall Lab)
FD300K65 |
300 kDa Starter Kit |
£204.00
Pack of 4 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
Description
- Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes
- Provides high recoveries, typically > 90%
- For research use only
Specifications
EFA: 15.2 cm2 | |
Non-sterile | |
4/Pk. | |
20 to 60 mL (Sample) |