missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Jumbosep™ Centrifugal Device Starter Kits
Jumbosep™ Centrifugal Device Starter Kit replaces dialysis, chemical precipitation, and lyophilization.
Brand: Cytiva FD300K65
This item is not returnable.
View return policy
Description
- Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes
- Provides high recoveries, typically > 90%
- For research use only
Specifications
Orange | |
PES | |
4/Pk. | |
20 to 60 mL (Sample) |
EFA: 15.2 cm2 | |
Non-sterile | |
300 kDa Starter Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Jumbosep™ Centrifugal Device Starter Kits > 300 kDa Starter Kit
Spot an opportunity for improvement?Share a Content Correction