Learn More
Justrite™ Biohazard Waste Can
The Justrite Biohazard Waste Can is a sturdy, steel can used to store contaminated laundry as well as other regulated waste. Can features a leakproof construction with foot-operated, self-closing lid.
Brand: Justrite™ 05930
Code : 03
Additional Details : Weight : 6.00000kg
Description
Safely segregate biohazard waste to ensure proper disposal of blood- or pathogen-contaminated laundry items and pathological and microbiological wastes.
- Features leakproof, sturdy steel construction with foot-operated, self-closing lid
- Eliminates any reason to reach into the waste area or touch the can
- Holds biohazard bag (sold separately) securely around circumference of can top
- Large biohazard label clearly identifies the contents and alerts workers
Not to be used for disposal of sharp items.
Specifications
FM, TUV, OSHA | |
14 in. | |
White | |
Foot-operated cover | |
18 in. | |
Biohazard Waste Container |
35.56 cm | |
10 gal. | |
35.5 cm | |
Safe, temporary collection of biological waste | |
Metal |