missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Biohazard Waste Can
The Justrite Biohazard Waste Can is a sturdy, steel can used to store contaminated laundry as well as other regulated waste. Can features a leakproof construction with foot-operated, self-closing lid.
£207.00 - £222.00
Specifications
Certifications/Compliance | FM, TUV, OSHA |
---|---|
Color | White |
For Use With (Application) | Safe, temporary collection of biological waste |
Material | Metal |
Product Type | Biohazard Waste Container |
Product Code | Brand | Capacity (English) | Diameter (English) Outer | Height (English) | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Diameter (English) Outer | Height (English) | Includes | Price | Quantity & Availability | ||
12373823
|
Justrite™
05930 |
10 gal. | 14 in. | 18 in. | Foot-operated cover |
£207.00
Each |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11934977
|
Justrite™
05910 |
6 gal. | 12 in. | 16 in. | Foot-operated cover |
£222.00
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12903685
|
Justrite™
05935 |
10 gal. | 13.94 in. | 18.25 in. | Foot-operated cover and soundguard |
£208.00
Each |
Estimated Shipment: 03-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Safely segregate biohazard waste to ensure proper disposal of blood- or pathogen-contaminated laundry items and pathological and microbiological wastes.
- Features leakproof, sturdy steel construction with foot-operated, self-closing lid
- Eliminates any reason to reach into the waste area or touch the can
- Holds biohazard bag (sold separately) securely around circumference of can top
- Large biohazard label clearly identifies the contents and alerts workers
Not to be used for disposal of sharp items.
Certifications
FM approved
Compliance
Complies with OSHA 29CFR1910.1030.
Specifications
FM, TUV, OSHA | |
Safe, temporary collection of biological waste | |
Biohazard Waste Container |
White | |
Metal |