Learn More
Justrite™ Horizontal and Vertical Drum Vents
For controlled release of flammable vapors; prevent explosions caused by pressure buildup
Brand:  Justrite™ 08300
 View more versions of this product
Product Code. 11943549
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Choose brass, stainless steel or nonmetallic vent
  • For petroleum-based liquids, use non-metallic or brass vents
  • For chlorinated solvents, use nonmetallic vent with stainless-steel flame arrester
  • For corrosives and general solvents, use stainless steel vents
Vertical Drum Vent:
  • Has operating valve that opens automatically to prevent pressure buildup inside drum
  • Horizontal Drum Vent
    • Has operating valve that can be opened manually to relieve internal pressure
    Order Info
    All vents fit 2 in. (5.1cm) bung. Cast Iron EL Fitting for use with 17-186A and 17-186B also available; see cat. no. 19018555.
    Certifications
    FM approved.
    Specifications
    Specifications
    FM
    Opens at 5 psig Internal Pressure
    Horizontal Drums
    Operating Valve
    Horizontal Drum Vent
    Controlled release of flammable vapors (Petroleum-based liquids), prevent explosions caused by pressure buildup
    Brass