missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Horizontal and Vertical Drum Vents
For controlled release of flammable vapors; prevent explosions caused by pressure buildup
£56.88 - £114.00
Specifications
Certifications/Compliance | FM |
---|---|
Description | Opens at 5 psig Internal Pressure |
Product Code | Brand | Product Type | For Use With (Application) | Includes | Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | For Use With (Application) | Includes | Material | Price | Quantity & Availability | ||
10279040
|
Justrite™
08101 |
Vertical Drum Vent | Controlled release of flammable vapors (Petroleum-based liquids), prevent explosions caused by pressure buildup | Operating Valve | Brass | N/A | |||
12716065
|
Justrite™
08005 |
Vertical Drum Vent | Controlled release of flammable vapors (Petroleum-based liquids), prevent explosions caused by pressure buildup | Operating Valve, Plated Steel Flame Arrester | Non-metallic Cast Iron |
£77.75
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
||
12726065
|
Justrite™
08006 |
Vertical Drum Vent | Controlled release of flammable vapors (Chlorinated solvents), prevent explosions caused by pressure buildup | Operating Valve, Stainless Steel Flame Arrester | Non-metallic Cast Iron |
£114.00
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
11943549
|
Justrite™
08300 |
Horizontal Drum Vent | Controlled release of flammable vapors (Petroleum-based liquids), prevent explosions caused by pressure buildup | Operating Valve | Brass | N/A | |||
Description
- Choose brass, stainless steel or nonmetallic vent
- For petroleum-based liquids, use non-metallic or brass vents
- For chlorinated solvents, use nonmetallic vent with stainless-steel flame arrester
- For corrosives and general solvents, use stainless steel vents
Vertical Drum Vent:
- Has operating valve that opens automatically to prevent pressure buildup inside drum
- Has operating valve that can be opened manually to relieve internal pressure
Order Info
All vents fit 2 in. (5.1cm) bung. Cast Iron EL Fitting for use with 17-186A and 17-186B also available; see cat. no. 19018555.
Certifications
FM approved.
Specifications
FM | |
Opens at 5 psig Internal Pressure |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title