missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Nonmetallic Dispensing Can

Permit easy, one-hand dispensing of hazardous liquids directly onto work surface. The nonmetallic dispensing can is made from polyethylene with a brass dispenser valve.

£46.37 - £64.00

Specifications

Product Type Dispensing Can
For Use With (Application) For use with Flammables
Material Polyethylene
Includes Leakproof Brass Dispenser Valves Self-close
View More Specs

Products 3
Product Code Brand Color Capacity (English) Capacity (Metric) Price Quantity & Availability  
Product Code Brand Color Capacity (English) Capacity (Metric) Price Quantity & Availability  
12901405
View Documents
Justrite™
14002
White 4 oz. 0.19 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11943679
View Documents
Justrite™
14010
Red 1 qt. 0.95 L
£64.00
Each
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
11933679
View Documents
Justrite™
14004
Red 0.5 pt. 0.24 L
£55.55
Each
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
Description

Description

  • Ideal for use with flammables
  • Leakproof, self-closing dispenser valves minimize spills, controls excess
  • High-density polyethylene body
  • Permits easy, one-hand dispensing of hazardous liquids directly onto work surface
Specifications

Specifications

Dispensing Can
For use with Flammables
Polyethylene
Leakproof Brass Dispenser Valves Self-close
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.