missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Nonmetallic Dispensing Can
Permit easy, one-hand dispensing of hazardous liquids directly onto work surface. The nonmetallic dispensing can is made from polyethylene with a brass dispenser valve.
Brand: Justrite™ 14002
This item is not returnable.
View return policy
Description
- Ideal for use with flammables
- Leakproof, self-closing dispenser valves minimize spills, controls excess
- High-density polyethylene body
- Permits easy, one-hand dispensing of hazardous liquids directly onto work surface
Specifications
Dispensing Can | |
0.19 L | |
For use with Flammables | |
Leakproof Brass Dispenser Valves Self-close | |
5.72 mm |
4 oz. | |
White | |
Polyethylene | |
2.25 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Justrite™ Nonmetallic Dispensing Can > 0.95L (1 qt.)
Spot an opportunity for improvement?Share a Content Correction