Learn More
Asecos™ K-PHOENIX-90 Combination Safety Storage Cabinets with Auto Close Doors
For the safe and separated storage of aggressive acids, alkalis and flammable hazardous substances in working areas.
Brand: Asecos™ 3033400133839
4149.29 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 500.00000kg
Description
- Hermetically sealed storage compartments with fire-resistant dividing walls
- Integrated aeration and venting system, corrosion resistance of the metal-free air ducts
- In accordance with EN 14470-1 and TRGS 510
- Robust construction and longevity
- Triple-hinged doors, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for protection against corrosion
- Door open arrest system as standard for the convenient and safe removal or storage of containers
- Doors lockable with profile cylinder with integration into an existing locking system possible
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- Fire resistance of 90 minutes
- Easy alignment – adjusting aids to compensate for uneven floors
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Some models with serial automatic door closing which releases arrested doors after approx. 60 seconds with an acoustic and visual signal before closing
Specifications
6 x 25 kg/5 L sheet steel drawers; 6 x 25 kg/11 L PP pull-out shelves with PP-sump | |
Storage of aggressive and flammable hazardous materials | |
Steel | |
617 x 1197 x 1969 mm | |
Light Gray | |
Wing | |
EN 14470-1, TRGS 510 |
Safety storage cabinet | |
Powder-coated | |
Door auto-close function | |
575 kg | |
6 x Drawers: 25 kg/5 L each, Pull-out shelves with sumps: 25 kg/11 L each, Max load: 600 kg | |
3 |