missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ K-PHOENIX-90 Combination Safety Storage Cabinets with Auto Close Doors
For the safe and separated storage of aggressive acids, alkalis and flammable hazardous substances in working areas.
£4845.26 - £4848.44
Specifications
Certifications/Compliance | EN 14470-1, TRGS 510 |
---|---|
Color | Light Gray |
Dimensions (D x W x H) | 617 x 1197 x 1969 mm |
Material | Steel |
Product Type | Safety storage cabinet |
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
15763738
|
Asecos™
3033400133839 |
6 x Drawers: 25 kg/5 L each, Pull-out shelves with sumps: 25 kg/11 L each, Max load: 600 kg |
£4848.44
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947941
|
Asecos™
3033600133842 |
600 kg |
£4845.26
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Hermetically sealed storage compartments with fire-resistant dividing walls
- Integrated aeration and venting system, corrosion resistance of the metal-free air ducts
- In accordance with EN 14470-1 and TRGS 510
- Robust construction and longevity
- Triple-hinged doors, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for protection against corrosion
- Door open arrest system as standard for the convenient and safe removal or storage of containers
- Doors lockable with profile cylinder with integration into an existing locking system possible
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- Fire resistance of 90 minutes
- Easy alignment – adjusting aids to compensate for uneven floors
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Some models with serial automatic door closing which releases arrested doors after approx. 60 seconds with an acoustic and visual signal before closing
Specifications
EN 14470-1, TRGS 510 | |
617 x 1197 x 1969 mm | |
Safety storage cabinet | |
Storage of aggressive and flammable hazardous materials | |
3 | |
575 kg |
Light Gray | |
Steel | |
Wing | |
Door auto-close function | |
Powder-coated |