missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Combi Safety Storage Cabinets K-UB-90
Safe and approved storage of aggressive and flammable hazardous substances in working areas
POA
Specifications
Depth (Metric) Exterior | 502 mm |
---|---|
Color | Light Gray |
Material | Steel |
Product Type | Combi safety storage cabinets |
Height (Metric) Exterior | 601 mm |
Alert:
For indoor use only
Product Code | Brand | Model | Weight (Metric) | No. of Doors | No. of Shelves | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Weight (Metric) | No. of Doors | No. of Shelves | Price | Quantity & Availability | ||
15914366
|
asecos™
3035800135362 |
K90.060.110.050.UB.ST | 152.50 kg | 1 | 1 |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |
15984366
|
asecos™
3037000135364 |
K90.060.140.050.UB.ST | 186.50 kg | 1 | 1 |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |
15411435
|
asecos™
3036500130366 |
K90.060.110.050.UB.2T | 152.50 kg | 2 | 1 |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |
15914376
|
asecos™
3037700133847 |
K90.060.140.050.UB.3T | 196.50 kg | 3 | 2 |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |
15184599
|
asecos™
3037700130378 |
K90.060.140.050.UB.3T | 190.50 kg | 3 | 1 |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Safe and approved storage of aggressive and flammable hazardous substances in working areas
Function / construction
- Robust construction and longevity: body made of powder-coated sheet steel
- User-friendly and safe: door and drawer remain open in any position, interior of the cabinet completely visible — access to all containers, 93° opening of the wing door
- Safe: drawer fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
- No unauthorised use: door and drawer lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system, extraction of both compartments through one ventilation connection
Certificates
- Fire resistance 90 minutes
- type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- CE compliant
Specifications
502 mm | |
Steel | |
601 mm | |
Powder-coated plain |
Light Gray | |
Combi safety storage cabinets | |
502 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
asecos™ Type 90 Combi Safety Storage Cabinets K-UB-90