missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Kanamycin sulfate Product Code.: 15274854

Thermo Scientific Chemicals Kanamycin sulfate

Product Code. 15274854
10 g
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy

Product Code. 15274854

Brand: Thermo Scientific Chemicals 450810100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kanamycin sulfate, CAS # 25389-94-0, is an aminoglycoside sulfate salt with antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Kanamycin sulfate is the sulfate salt form of kanamycin A
  • It was isolated from Streptomyces kanamyceticus
  • This compound inhibits translocation during protein biosynthesis. It binds to the 30S ribosomal subunit, causing misreading of the mRNA sequence

Application

  • Kanamycin sulfate shows antibacterial activity. It is effective against gram-positive and gram-negative bacteria, as well as Mycoplasma species
  • This compound is also involved in translating prokaryotic ribosomes
  • In molecular biology experiments, it is commonly used to select and isolate transformants expressing the kanamycin resistance gene, and plants that have the NPT II (APH3) gene incorporated
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25389-94-0
Molecular Formula C18H38N4O15S
Molecular Weight (g/mol) 582.575
MDL Number MFCD00070253
InChI Key OOYGSFOGFJDDHP-IZQIRFRQSA-N
PubChem CID 134129479
IUPAC Name (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acidShow MoreShow Less
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O

Specifications

Type Kanamycin Sulfate
Packaging Glass Bottle
Quantity 10 g
Solubility Information Solubility in water: soluble.
Formula Weight 582.59
Chemical Name or Material Kanamycin sulfate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0454231 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number450810100,450810500,450811000
Lot NumberA0459576 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog Number450810100,450810500,450811000
Lot NumberA0454231 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number450810100,450810500,450811000
Lot NumberA0447974 Certificate TypeCertificate of Analysis Date09/12/2022 Catalog Number450810100,450810500,450811000
Lot NumberA0443484 Certificate TypeCertificate of Analysis Date22/06/2022 Catalog Number450810100,450810500,450811000
compliance-icons
Product Identifier
  • Kanamycin Sulfate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Kanamycin sulfate >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.