missing translation for 'onlineSavingsMsg'
Learn More
Kanamycin Sulfate, Ultrapure, Thermo Scientific Chemicals Product Code.: 15815168

Kanamycin Sulfate, Ultrapure, Thermo Scientific Chemicals

Product Code. 15815168 Shop All Thermo Scientific Products
5 g, Plastic Bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 15815168

Brand: Thermo Scientific J17924.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A selectable marker for cosmid vectors used in transfection of mammalian cells.

Mode of Action: Binds to ribosomal components and inhibits protein synthesis. Kanamycin is inactivated by an aminoglycoside phospho transferase (MW 25,000) in the periplasmic space.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selectable marker for cosmid vectors used in transfection of mammalian cells
  • Mode of Action: Binds to ribosomal components and inhibits protein synthesis
  • Kanamycin is inactivated by aminoglycoside phospho transferase (MW 25,000) in
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25389-94-0
Molecular Formula C18H38N4O15S
Molecular Weight (g/mol) 582.575
MDL Number MFCD00070253
InChI Key OOYGSFOGFJDDHP-IZQIRFRQSA-N
Synonym Kanamycin A
PubChem CID 134129479
IUPAC Name (2 R,3 S,4 S,5 S,6 R)-2-(aminomethyl)-6-[(1 R,2 S,3 S,4 R,6 S)-4,6-diamino-3-[(2 S,3 R,4 S,5 R,6 R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acidShow MoreShow Less
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O

Specifications

Type Kanamycin Sulfate
Physical Form Powder
Packaging Plastic Bottle
Quantity 5 g
Formula Weight 582.58
Chemical Name or Material Kanamycin sulfate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number241673 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ17924.14,J17924.06
Lot Number235389 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ17924.14,J17924.06
Lot Number235841 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ17924.14,J17924.06
Lot Number197845 Certificate TypeCertificate of Analysis Date15/11/2023 Catalog NumberJ17924.14,J17924.06
Lot Number198328 Certificate TypeCertificate of Analysis Date15/11/2023 Catalog NumberJ17924.14,J17924.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kanamycin Sulfate, Ultrapure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.