missing translation for 'onlineSavingsMsg'
Learn More
Kaolin, pure, Thermo Scientific Chemicals Product Code.: 11984321

Kaolin, pure, Thermo Scientific Chemicals

Product Code. 11984321
10 kg, Plastic drum
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 11984321

Brand: Thermo Scientific Acros 211740100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1332-58-7
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
MDL Number MFCD00062311
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
Synonym kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
PubChem CID 56841936
IUPAC Name oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O

Specifications

Melting Point 1750.0°C
Color Beige
Physical Form Powder
Quantity 10 kg
Merck Index 14,5282
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in cold acids and alkali hydroxides
Formula Weight 258.2
Grade Pure
Loss on Drying 1.5% max.
Loss on Ignition 10.8% to 11.8%
Packaging Plastic drum
Chemical Name or Material Kaolin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465235 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog Number211740025,211740100,211740010
Lot NumberA0459927 Certificate TypeCertificate of Analysis Date08/02/2024 Catalog Number211740025,211740100,211740010
Lot NumberA0456188 Certificate TypeCertificate of Analysis Date15/09/2023 Catalog Number211740025,211740100,211740010
Lot NumberA0454625 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number211740025,211740100,211740010
Lot NumberA0449731 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog Number211740025,211740100,211740010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kaolin, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.