Learn More
DWK Life Sciences KECK™ Clip for Spherical Joints, POM, S
KECK™ spherical ground joints ensure a secure connection.
Brand: DWK Life Sciences
Additional Details : Weight : 1.00000kg
Description
KECK™ spherical ground joint clamps represent a favourable alternative to conventional spherical ground joint clamps made of metal. They are easy to put on and take off. The good spring properties of the POM material prevent scratching of the glass surface. An important contribution to safety in the laboratory. The different colours make the sizes easy to distinguish from each other.
- Excellent dimensional stability and good spring properties
- No damage to the glass surface during attachment unlike conventional wire clamps
- Easy, safe and quick to put on and take off
- Colour coding per size
- Cost-effective alternative to conventional spherical ground joint clips made of metal
Specifications
100 | |
Securing of Spherical Ground Joints | |
Light Blue |
POM | |
Securing of spherical ground joints |