Learn More
DWK Life Sciences™ KECK™ Tubing Clamp, PBT, KT
For infinitely variable regulation of flow rates.
Brand: DWK Life Sciences™ 286338204
This item is not returnable.
View return policy
Description
With just one finger, flow rates in hoses can be infinitely adjusted with KECK™ hose clamps. The clamps are available in different colours for the different hose diameters.
- Continuous regulation of flow rates
- Handling with one hand possible
- Color coding of the clamps according to the different hose diameters
Specifications
100 | |
Infinitely Variable Regulation of Flow Rates | |
Fits tube OD 6 mm |
PBT | |
infinitely variable regulation of flow rates | |
Yellow |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
DWK Life Sciences™ KECK™ Tubing Clamp, PBT, KT > Fits tube OD 6 mm
Spot an opportunity for improvement?Share a Content Correction