Learn More
Keratinocyte-SFM Medium (Kit) with L-glutamine, EGF, and BPE
GREENER_CHOICE
Brand:  Gibco 17005075
Product Code. 10144892
  • £132.00 / 1 set
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
with L-Glutamine, EGF and BPE.
Shelf life after supplementation: 2 weeks
TRUSTED_SUSTAINABILITY
Specifications
Specifications
• 500 ml 1 X Keratinocyte-SFM with L-glutamine (store at 2°C to 8°C)
• 1 vial Human Recombinant EGF (store at -5°C to -30°C)
• 1 vial Bovine Pituitary Extract (store at -5°C to -30°C)
Serum-free
Serum Free Medium (SFM)
Sterile
Glutamine, Phenol Red, EGF, BPE
1 Kit
Keratinocyte Cells, Epithelial Cells
Mammalian Cell Culture
Serum-free
Sterile-filtered
7 to 8.1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Keratinocyte-SFM Medium (Kit) with L-glutamine, EGF, and BPE >