missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, Reagent grade, Honeywell™ Riedel-de Haën™

reagent grade, low odor

£94.02 - £513.67

Chemical Identifiers

CAS 8008-20-6
MDL Number MFCD00135561
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15693860
View Documents
Honeywell Research Chemicals
329460-3.8L
3.8 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15683860
View Documents
Honeywell Research Chemicals
329460-18.9L
18.9 L Stainless Steel Can
£513.67
18.90 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

8008-20-6
MFCD00135561

Specifications

8008-20-6
0.8g/mL at 25°C
70°C (158°F)
UN1223
Reagent
Kerosene
Colorless
175°C to 325°C
MFCD00135561
4.5 (vs air)
Liquid
Videos
Safety and Handling

Safety and Handling

P301 + P310-P331

H304-H315-H336-H411

Danger

missing translation for 'dotInformation' : class 3 - PG 3 - DOT_1993NA_ Kerosene

missing translation for 'einecsNumber' : 232-366-4

missing translation for 'rtecsNumber' : OA5500000

SDS
Documents

Documents

Product Certifications