Learn More
Product Code | Brand | Product Type | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | For Use With (Equipment) | Price | Quantity & Availability | |||||
16552452
|
Thermo Scientific™
97002580 |
24 Combi tip comb | KingFisher Apex |
£764.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12653116
|
Thermo Scientific™
95040470 |
24 Deep Well plate, sterile | KF Duo Prime, Flex, Presto |
£203.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12663116
|
Thermo Scientific™
95040480 |
24 Deep Well plate | KF Duo Prime, Flex, Presto |
£251.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12624075
|
Thermo Scientific™
97002610 |
24 DW tip comb and plate | KingFisher Flex and Presto |
£576.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12643116
|
Thermo Scientific™
97003510 |
6-tip comb and 24 DW plate | KingFisher Duo Prime |
£179.00
Pack of 48 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17140069
|
Thermo Scientific™
95040470B |
24 Deep Well plate, barcoded | KingFisher Apex |
£214.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17190069
|
Thermo Scientific™
97002610B |
24 DW tip comb, plate, barcode | KingFisher Apex |
£600.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17150069
|
Thermo Scientific™
95040480B |
24 DW plate, barcoded, sterile | KingFisher Apex |
£267.00
Pack of 50 |
Estimated Shipment: 29-05-2025
Log in to see stock. |
|||||
17100079
|
Thermo Scientific™
97002620B |
24 DW tip comb, plate, sterile | KingFisher Apex |
£620.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.