missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kieselguhr, Pure, Washed with Acid, Fisher Chemical™
£114.72 - £315.00
Specifications
Color | Gray |
---|---|
pH | 9 to 10 |
Physical Form | Solid |
CAS | 61790-53-2 |
Molecular Formula | O2Si |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10385850
|
Fisher Chemical
K/0100/53 |
500 g | Plastic Powder Jar | N/A | N/A | N/A | |||||
10763991
|
Fisher Chemical
K/0100/60 |
1 kg | Plastic Keg |
£315.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Specifications
Gray | |
Solid | |
O2Si |
9 to 10 | |
61790-53-2 | |
132803 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Kieselguhr
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity after repeated exposure Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H373-May cause damage to organs.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kieselguhr, Pure, Washed with Acid, Fisher Chemical™